3-Bromo-4-chloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

55930-1G 55930-500MG 55930-5G
76917-222EA 500.75 USD
76917-222 76917-224 76917-226
3-Bromo-4-chloro-2-fluorobenzoic acid ≥97%
3-Bromo-4-chloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28398041
CAS Number: 1677706-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR