3-Isopropoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
55685-25G
55685-5G
76917-210EA
460.44
USD
76917-210
76917-212
3-Isopropoxybenzaldehyde ≥97%
3-Isopropoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01922005 CAS Number: 75792-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...