3-Bromo-2-iodo-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

54999-1G 54999-25G 54999-500MG 54999-5G
76917-180EA 218.55 USD
76917-180 76917-182 76917-184 76917-186
3-Bromo-2-iodo-5-methylbenzoic acid ≥95%
3-Bromo-2-iodo-5-methylbenzoic acid
Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20484515
CAS Number: 750585-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR