4-Bromo-1-(2-methoxyethyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

54811-1G 54811-25G 54811-5G
76917-168EA 62.6 USD
76917-168 76917-170 76917-172
4-Bromo-1-(2-methoxyethyl)-1H-pyrazole ≥97%
4-Bromo-1-(2-methoxyethyl)-1H-pyrazole
Formula: C₆H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD12405645
CAS Number: 847818-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR