3,5-Difluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

54684-1G 54684-25G 54684-500MG 54684-5G
76917-152EA 147.47 USD
76917-152 76917-154 76917-156 76917-158
3,5-Difluoro-N-methylbenzamide ≥97%
3,5-Difluoro-N-methylbenzamide
Formula: C₈H₇F₂NO
MW: 171.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19595536
CAS Number: 948710-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR