3-Amino-5-bromobenzamide ≥95% research grade

Supplier: AOBChem USA

Warning

54279-1G 54279-25G 54279-500MG 54279-5G
76917-116EA 236.59 USD
76917-116 76917-118 76917-120 76917-122
3-Amino-5-bromobenzamide ≥95% research grade
3-Amino-5-bromobenzamide
Formula: C₇H₇BrN₂O
MW: 215.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18389345
CAS Number: 1261609-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR