3-Bromo-5-chloro-2,6-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

53851-1G 53851-25G 53851-500MG 53851-5G
76917-084EA 183.54 USD
76917-084 76917-086 76917-088 76917-090
3-Bromo-5-chloro-2,6-difluorobenzaldehyde ≥95%
3-Bromo-5-chloro-2,6-difluorobenzaldehyde
Formula: C₇H₂BrClF₂O
Storage Temperature: Ambient
MDL Number: MFCD12545924
CAS Number: 2514963-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR