4-Bromo-1-(1,1-difluoroethyl)-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

53463-1G 53463-500MG 53463-5G
76917-052EA 524.09 USD
76917-052 76917-054 76917-056
4-Bromo-1-(1,1-difluoroethyl)-2-fluorobenzene ≥95%
4-Bromo-1-(1,1-difluoroethyl)-2-fluorobenzene
Formula: C₈H₆BrF₃
Storage Temperature: Ambient
MDL Number: MFCD21602847
CAS Number: 1823231-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR