3-Iodo-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

53441-1G 53441-25G 53441-500MG 53441-5G
76917-036EA 379.81 USD
76917-036 76917-038 76917-040 76917-042
3-Iodo-5-phenylpyridine ≥97%
3-Iodo-5-phenylpyridine
Formula: C₁₁H₈IN
Storage Temperature: Refrigerator
MDL Number: MFCD14702052
CAS Number: 1214362-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR