4-Bromo-2-chloro-5-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

53414-1G 53414-25G 53414-5G
76917-024EA 62.6 USD
76917-024 76917-026 76917-028
4-Bromo-2-chloro-5-methylpyridine ≥97% research grade
4-Bromo-2-chloro-5-methylpyridine
Formula: C₆H₅BrClN
Storage Temperature: Refrigerator
MDL Number: MFCD11100650
CAS Number: 867279-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR