3-Fluoro-2-formyl-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53132-1G 53132-500MG 53132-5G
76916-994EA 589.87 USD
76916-994 76916-996 76916-998
3-Fluoro-2-formyl-4-methylbenzoic acid ≥97%
3-Fluoro-2-formyl-4-methylbenzoic acid
Formula: C₉H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD34178671
CAS Number: 2382988-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR