3-Fluoro-5-iodophenol ≥97% research grade

Supplier: AOBChem USA

Warning

53110-1G 53110-25G 53110-500MG 53110-5G
76916-978EA 173.99 USD
76916-978 76916-980 76916-982 76916-984
3-Fluoro-5-iodophenol ≥97% research grade
3-Fluoro-5-iodophenol
Formula: C₆H₄FIO
Storage Temperature: Refrigerator
MDL Number: MFCD16999911
CAS Number: 939771-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR