3-Bromo-[1,1'-biphenyl]-2-ol ≥97%

Supplier: AOBChem USA

Warning

53073-25G 53073-5G
76916-972EA 204.76 USD
76916-972 76916-974
3-Bromo-[1,1'-biphenyl]-2-ol ≥97%
3-Bromo-[1,1'-biphenyl]-2-ol
Formula: C₁₂H₉BrO
MW: 249.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21362356
CAS Number: 23197-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR