3-Bromo-5-chloro-4-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

52978-1G 52978-25G 52978-500MG 52978-5G
76916-928EA 436.03 USD
76916-928 76916-930 76916-932 76916-934
3-Bromo-5-chloro-4-fluorobenzoic acid ≥95%
3-Bromo-5-chloro-4-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD24322471
CAS Number: 176548-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR