3-Ethoxy-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

52904-1G 52904-25G 52904-500MG 52904-5G
76916-918EA 168.69 USD
76916-918 76916-920 76916-922 76916-924
3-Ethoxy-5-methylbenzoic acid ≥95%
3-Ethoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27940681
CAS Number: 500901-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR