3',4'-Dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

52731-1G 52731-500MG 52731-5G
76916-906EA 498.63 USD
76916-906 76916-908 76916-910
3',4'-Dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%
3',4'-Dimethoxy-[1,1'-biphenyl]-4-carbaldehyde
Formula: C₁₅H₁₄O₃
MW: 242.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424648
CAS Number: 640769-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR