3-Formyl-5-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52498-1G 52498-25G 52498-500MG 52498-5G
76916-886EA 292.81 USD
76916-886 76916-888 76916-890 76916-892
3-Formyl-5-isopropylbenzoic acid ≥97%
3-Formyl-5-isopropylbenzoic acid
Formula: C₁₁H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD20660434
CAS Number: 1187028-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR