3-Fluoro-6-methoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52089-1G 52089-500MG 52089-5G
76916-838EA 498.63 USD
76916-838 76916-840 76916-842
3-Fluoro-6-methoxy-2-methylbenzaldehyde ≥97%
3-Fluoro-6-methoxy-2-methylbenzaldehyde
Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD28647479
CAS Number: 1781868-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR