3-Chloro-4-methylpicolinic acid ≥95%

Supplier: AOBChem USA

Warning

51808-1G 51808-500MG 51808-5G
76916-814EA 647.15 USD
76916-814 76916-816 76916-818
3-Chloro-4-methylpicolinic acid ≥95%
3-Chloro-4-methylpicolinic acid
Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257432
CAS Number: 1211518-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR