3-(Difluoromethoxy)-4-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

51191-1G 51191-25G 51191-500MG 51191-5G
76916-788EA 241.89 USD
76916-788 76916-790 76916-792 76916-794
3-(Difluoromethoxy)-4-fluorobenzaldehyde ≥97% research grade
3-(Difluoromethoxy)-4-fluorobenzaldehyde
Formula: C₈H₅F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698501
CAS Number: 1214367-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR