3-Chloro-2,6-difluoroisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

49705-1G 49705-500MG 49705-5G
76916-710EA 589.87 USD
76916-710 76916-712 76916-714
3-Chloro-2,6-difluoroisonicotinaldehyde ≥97%
3-Chloro-2,6-difluoroisonicotinaldehyde
Formula: C₆H₂ClF₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD32874815
CAS Number: 2551116-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR