4-Bromo-1-(difluoromethoxy)-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49619-1G 49619-25G 49619-500MG 49619-5G
76916-696EA 439.22 USD
76916-696 76916-698 76916-700 76916-702
4-Bromo-1-(difluoromethoxy)-2-(trifluoromethyl)benzene ≥97%
4-Bromo-1-(difluoromethoxy)-2-(trifluoromethyl)benzene
Formula: C₈H₄BrF₅O
MW: 291.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036076
CAS Number: 954236-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR