3-Chloro-2,4-dimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

49348-1G 49348-25G 49348-500MG 49348-5G
76916-666EA 183.54 USD
76916-666 76916-668 76916-670 76916-672
3-Chloro-2,4-dimethoxybenzaldehyde ≥95%
3-Chloro-2,4-dimethoxybenzaldehyde
Formula: C₉H₉ClO₃
MW: 200.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835191
CAS Number: 72482-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR