3-Bromo-2-fluoro-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48877-1G 48877-25G 48877-500MG 48877-5G
76916-640EA 168.69 USD
76916-640 76916-642 76916-644 76916-646
3-Bromo-2-fluoro-5-iodobenzaldehyde ≥97%
3-Bromo-2-fluoro-5-iodobenzaldehyde
Formula: C₇H₃BrFIO
Storage Temperature: Refrigerator
MDL Number: MFCD32856723
CAS Number: 2091143-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR