3-Bromo-2-fluoro-6-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

48818-1G 48818-25G 48818-5G
76916-626EA 88.06 USD
76916-626 76916-628 76916-630
3-Bromo-2-fluoro-6-(trifluoromethyl)benzoic acid ≥97%
3-Bromo-2-fluoro-6-(trifluoromethyl)benzoic acid
Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29049268
CAS Number: 1980062-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR