3-Bromo-2-fluoro-6-methyl-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-fluoro-6-methylbenzoic acid

48748-1G 48748-500MG 48748-5G
76916-616EA 587.74 USD
76916-616 76916-618 76916-620
3-Bromo-2-fluoro-6-methyl-benzoic acid ≥97%
3-Bromo-2-fluoro-6-methyl-benzoic acid
Formula: C₈H₆BrFO₂
MW: 233.0345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712077
CAS Number: 1427433-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR