3,5-Dichloro-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

48621-1G 48621-500MG 48621-5G
76916-604EA 855.09 USD
76916-604 76916-606 76916-608
3,5-Dichloro-2-methoxyphenylboronic acid ≥97%
3,5-Dichloro-2-methoxyphenylboronic acid
Formula: C₇H₇BCl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18072762
CAS Number: 2573074-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR