3-(Benzyloxy)-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

47726-1G 47726-25G 47726-500MG 47726-5G
76916-532EA 206.88 USD
76916-532 76916-534 76916-536 76916-538
3-(Benzyloxy)-2-methylphenylboronic acid ≥97%
3-(Benzyloxy)-2-methylphenylboronic acid
Formula: C₁₄H₁₅BO₃
MW: 242.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31543955
CAS Number: 2377609-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR