3-Chloro-2-fluoro-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

47271-1G 47271-25G 47271-5G
76916-504EA 64.72 USD
76916-504 76916-506 76916-508
3-Chloro-2-fluoro-5-iodobenzaldehyde ≥97%
3-Chloro-2-fluoro-5-iodobenzaldehyde
Formula: C₇H₃ClFIO
MW: 284.454 g/mol
MDL Number: MFCD32902706
CAS Number: 2304494-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR