3-Bromo-5-(tert-butyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47140-1G 47140-25G 47140-500MG 47140-5G
76916-488EA 247.19 USD
76916-488 76916-490 76916-492 76916-494
3-Bromo-5-(tert-butyl)benzaldehyde ≥97%
3-Bromo-5-(tert-butyl)benzaldehyde
Formula: C₁₁H₁₃BrO
Storage Temperature: Refrigerator
MDL Number: MFCD27947907
CAS Number: 241155-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR