4-(Phenoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46926-1G 46926-25G 46926-500MG 46926-5G
76916-474EA 271.6 USD
76916-474 76916-476 76916-478 76916-480
4-(Phenoxymethyl)benzaldehyde ≥97%
4-(Phenoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09879939
CAS Number: 2683-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR