3-Bromofuran-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

46627-1G 46627-25G 46627-500MG 46627-5G
76916-458EA 147.47 USD
76916-458 76916-460 76916-462 76916-464
3-Bromofuran-2-carboxylic acid ≥97%
3-Bromofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₃
MW: 190.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00092761
CAS Number: 14903-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR