3-Fluoro-4-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46339-1G 46339-25G 46339-500MG 46339-5G
76916-442EA 248.26 USD
76916-442 76916-444 76916-446 76916-448
3-Fluoro-4-propoxybenzaldehyde ≥97%
3-Fluoro-4-propoxybenzaldehyde
Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD06247716
CAS Number: 19415-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR