3-(Ethylcarbamoyl)-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

46104-1G 46104-25G 46104-5G
76916-430EA 66.84 USD
76916-430 76916-432 76916-434
3-(Ethylcarbamoyl)-2-fluorophenylboronic acid ≥97%
3-(Ethylcarbamoyl)-2-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
MW: 210.998 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22682879
CAS Number: 2096353-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR