3-Bromo-5-fluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

45944-1G 45944-25G 45944-5G
76916-404EA 70.02 USD
76916-404 76916-406 76916-408
3-Bromo-5-fluoro-N-methylbenzamide ≥97%
3-Bromo-5-fluoro-N-methylbenzamide
Formula: C₈H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD13399077
CAS Number: 1211635-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR