4-(5-Bromopyrimidin-2-yl)thiomorpholine ≥97%

Supplier: AOBChem USA

Warning

45670-25G 45670-5G
76916-378EA 240.83 USD
76916-378 76916-380
4-(5-Bromopyrimidin-2-yl)thiomorpholine ≥97%
4-(5-Bromopyrimidin-2-yl)thiomorpholine
Formula: C₈H₁₀BrN₃S
MW: 260.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909661
CAS Number: 1341761-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR