3-Bromo-2,4-dichloro-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

45527-1G 45527-25G 45527-500MG 45527-5G
76916-342EA 173.99 USD
76916-342 76916-344 76916-346 76916-348
3-Bromo-2,4-dichloro-6-fluorobenzaldehyde ≥97%
3-Bromo-2,4-dichloro-6-fluorobenzaldehyde
Formula: C₇H₂BrCl₂FO
MW: 271.899 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32902676
CAS Number: 2586125-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR