3,5-Dibromo-2-chloro-6-methoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

45421-25G 45421-5G
76916-318EA 346.92 USD
76916-318 76916-320
3,5-Dibromo-2-chloro-6-methoxypyridine ≥97% research grade
3,5-Dibromo-2-chloro-6-methoxypyridine
Formula: C₆H₄Br₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD32707116
CAS Number: 2432849-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR