3-Bromo-5-chloro-4-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

44860-1G 44860-25G 44860-500MG 44860-5G
76916-290EA 274.78 USD
76916-290 76916-292 76916-294 76916-296
3-Bromo-5-chloro-4-fluorophenylboronic acid ≥95%
3-Bromo-5-chloro-4-fluorophenylboronic acid
Formula: C₆H₄BBrClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD29043325
CAS Number: 2377608-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR