2-(3-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-(methylthio)phenylboronic acid pinacol ester

44663-1G 44663-25G 44663-500MG 44663-5G
76916-276EA 177.18 USD
76916-276 76916-278 76916-280 76916-282
2-(3-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(3-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₈BFO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD22494876
CAS Number: 1351499-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR