3-Isopropoxy-4-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40912-1G 40912-25G 40912-500MG 40912-5G
76916-214EA 381.93 USD
76916-214 76916-216 76916-218 76916-220
3-Isopropoxy-4-(trifluoromethoxy)phenylboronic acid ≥97%
3-Isopropoxy-4-(trifluoromethoxy)phenylboronic acid
Formula: C₁₀H₁₂BF₃O₄
MW: 264.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21332936
CAS Number: 1793003-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR