3-Chloro-4-fluoro-5-(2-methylpropoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40888-1G 40888-25G 40888-5G
76916-200EA 76.39 USD
76916-200 76916-202 76916-204
3-Chloro-4-fluoro-5-(2-methylpropoxy)phenylboronic acid ≥97%
3-Chloro-4-fluoro-5-(2-methylpropoxy)phenylboronic acid
Formula: C₁₀H₁₃BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD20529449
CAS Number: 1793003-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR