(3-Bromo-2-chlorophenyl)boronic acid ≥95% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Bromo-2-chlorophenylboronic acid
40750-1G
40750-25G
40750-500MG
40750-5G
76916-186EA
160.2
USD
76916-186
76916-188
76916-190
76916-192
(3-Bromo-2-chlorophenyl)boronic acid ≥95% research grade
(3-Bromo-2-chlorophenyl)boronic acid
Formula:
C₆H₅BBrClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13184263 CAS Number: 352535-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...