2-(4-(Benzyloxy)-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-3-methylphenylboronic acid pinacol ester

40531-1G 40531-25G 40531-5G
76916-174EA 99.73 USD
76916-174 76916-176 76916-178
2-(4-(Benzyloxy)-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Benzyloxy)-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₂₀H₂₅BO₃
MW: 324.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383421
CAS Number: 1257078-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR