3-Fluoro-5-formylbenzeneboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, Benzaldehyde, 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 3-Fluoro-5-formylphenylboronic acid pinacol ester
40270-1G
40270-25G
40270-500MG
40270-5G
76916-132EA
183.54
USD
76916-132
76916-134
76916-136
76916-138
3-Fluoro-5-formylbenzeneboronic acid pinacol ester ≥97%
3-Fluoro-5-formylbenzeneboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BFO₃ MW: 250.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16996301 CAS Number: 1112209-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...