3-Ethoxy-2,4-difluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

40218-1G 40218-25G 40218-5G
76916-118EA 96.55 USD
76916-118 76916-120 76916-122
3-Ethoxy-2,4-difluorophenylboronic acid ≥97% research grade
3-Ethoxy-2,4-difluorophenylboronic acid
Formula: C₈H₉BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD19981528
CAS Number: 2096341-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR