3-Ethoxy-5-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

39750-1G 39750-25G 39750-5G
76916-074EA 94.43 USD
76916-074 76916-076 76916-078
3-Ethoxy-5-methylphenylboronic acid ≥97% research grade
3-Ethoxy-5-methylphenylboronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16295066
CAS Number: 1256346-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR