(3-(Cyclopropylmethoxy)-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-(Cyclopropylmethoxy)-5-methylphenylboronic acid

39714-1G 39714-25G 39714-5G 39714-500MG
76916-062EA 102.91 USD
76916-062 76916-064 76916-066 77650-572
(3-(Cyclopropylmethoxy)-5-methylphenyl)boronic acid ≥97% research grade
(3-(Cyclopropylmethoxy)-5-methylphenyl)boronic acid
Formula: C₁₁H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD17015733
CAS Number: 1256345-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR