3,6-Dibromo-2-methoxypyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
38831-1G
38831-25G
38831-500MG
38831-5G
76916-008EA
338.43
USD
76916-008
76916-010
76916-012
76916-014
3,6-Dibromo-2-methoxypyridine ≥97%
3,6-Dibromo-2-methoxypyridine
Formula:
C₆H₅Br₂NO MW: 266.918 g/mol Storage Temperature: Ambient |
CAS Number:
1806328-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...