3-Bromo-5-chloro-2-methoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

38605-1G 38605-500MG 38605-5G
76915-978EA 503.93 USD
76915-978 76915-980 76915-982
3-Bromo-5-chloro-2-methoxybenzoic acid ≥97% research grade
3-Bromo-5-chloro-2-methoxybenzoic acid
Formula: C₈H₆BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD08444538
CAS Number: 66237-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR